Gene Transcripts

Gene Symbol
OR5L2
Entrez Gene ID
26338
Gene Name
olfactory receptor family 5 subfamily L member 2
Gene Aliases
HSHTPCRX16, HTPCRX16, OR11-153
Chromosome Location
Chr. 11: 55827219 - 55828154 on Build GRCh38
UniGene ID
Hs.528356
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004739.1 NP_001004739.1 1 483
GenBank mRNA X64990.1 - 1 109

View Full Product Details