Gene Transcripts

Gene Symbol
MYEOV
Entrez Gene ID
26579
Gene Name
myeloma overexpressed
Gene Aliases
OCIM
Chromosome Location
Chr. 11: 69294138 - 69297287 on Build GRCh38
UniGene ID
Hs.523848
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293291.1 NP_001280220.1 3 2355
NM_001293294.1 NP_001280223.1 2 2451
NM_001293296.1 NP_001280225.1 2 2283
NM_001300923.1 NP_001287852.1 3 2187
NM_001300924.1 NP_001287853.1 2 2092
NM_138768.3 NP_620123.2 3 2261
GenBank mRNA AJ223366.1 - 2 2429
AK026148.1 - 2 2263
BC011815.2 - 3 2219 3163949

View Full Product Details