Gene Transcripts

Gene Symbol
OR8B2
Entrez Gene ID
26595
Gene Name
olfactory receptor family 8 subfamily B member 2
Gene Aliases
OR11-309, OR11-310
Chromosome Location
Chr. 11: 124382402 - 124397319 on Build GRCh38
UniGene ID
Hs.576944
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005468.1 NP_001005468.1 1 580
XM_017017535.1 XP_016873024.1 3 812
XM_017017536.1 XP_016873025.1 3 798

Gene Symbol
OR8B3
Entrez Gene ID
390271
Gene Name
olfactory receptor family 8 subfamily B member 3
Gene Aliases
OR11-311
Chromosome Location
Chr. 11: 124396410 - 124410495 on Build GRCh38
UniGene ID
Hs.690456
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005467.1 NP_001005467.1 1 580
XM_017017716.1 XP_016873205.1 6 1946
XM_017017717.1 XP_016873206.1 6 1249
XM_017017718.1 XP_016873207.1 4 1677

View Full Product Details