Gene Transcripts

Gene Symbol
OR7C1
Entrez Gene ID
26664
Gene Name
olfactory receptor family 7 subfamily C member 1
Gene Aliases
CIT-HSP-146E8, HSTPCR86P, OR19-5, OR7C4, TPCR86
Chromosome Location
Chr. 19: 14797550 - 14835376 on Build GRCh38
UniGene ID
Hs.466053
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198944.1 NP_945182.1 1 509
XM_005259853.3 XP_005259910.1 3 1013
XM_011527915.2 XP_011526217.1 5 1434

View Full Product Details