Gene Transcripts

Gene Symbol
OR2J2
Entrez Gene ID
26707
Gene Name
olfactory receptor family 2 subfamily J member 2
Gene Aliases
OR6-19, OR6-8, OR6.3.8, ORL684, dJ80I19.4, hs6M1-6
Chromosome Location
Chr. 6: 29173534 - 29174574 on Build GRCh38
UniGene ID
Hs.163518
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030905.2 NP_112167.2 1 559
GenBank mRNA BC069121.1 - 1 457
BC108896.1 - 1 460 40003949
BC108897.1 - 1 457 40003955
BC108898.1 - 1 457 40003956
BC108899.1 - 1 457 40003957
BU171389.1 - 1 561

View Full Product Details