Gene Transcripts

Gene Symbol
OR1L3
Entrez Gene ID
26735
Gene Name
olfactory receptor family 1 subfamily L member 3
Gene Aliases
OR9-28, OR9-D
Chromosome Location
Chr. 9: 122675130 - 122676104 on Build GRCh38
UniGene ID
Hs.626839
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005234.1 NP_001005234.1 1 636
GenBank mRNA BC136857.1 - 1 730
BC136859.1 - 1 730 9020850

View Full Product Details