Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015692.2 | NP_056507.2 | 30 | 4109 | |
XM_011527917.1 | XP_011526219.1 | 30 | 4076 | ||
XM_011527918.1 | XP_011526220.1 | 30 | 4112 | ||
XM_011527919.1 | XP_011526221.1 | 30 | 4112 | ||
XM_011527920.2 | XP_011526222.1 | 30 | 4112 | ||
XM_011527921.2 | XP_011526223.1 | 30 | 4112 | ||
XM_011527922.1 | XP_011526224.1 | 30 | 4112 | ||
XM_011527923.1 | XP_011526225.1 | 30 | 4112 | ||
XM_011527924.2 | XP_011526226.1 | 30 | 4112 | ||
XM_011527925.1 | XP_011526227.1 | 14 | 1954 | ||
XM_017026594.1 | XP_016882083.1 | 30 | 4112 | ||
GenBank mRNA | AB033109.1 | - | 30 | 3934 | |
AY101765.1 | - | 30 | 3953 |