Gene Transcripts

Gene Symbol
BRPF3
Entrez Gene ID
27154
Gene Name
bromodomain and PHD finger containing 3
Gene Aliases
Chromosome Location
Chr. 6: 36196773 - 36232790 on Build GRCh38
UniGene ID
Hs.520096
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015695.2 NP_056510.2 2 439
XM_005249010.2 XP_005249067.1 2 321
XM_005249011.3 XP_005249068.1 2 445
XM_011514489.1 XP_011512791.1 2 343
XM_011514490.1 XP_011512792.1 1 241
XM_011514491.1 XP_011512793.1 1 241
XM_011514492.1 XP_011512794.1 1 241
XM_017010742.1 XP_016866231.1 1 241
GenBank mRNA AB033112.1 - 2 439
BC033652.2 - 1 143
BC117387.1 - 1 246 40125938
BC143917.1 - 1 246 9052435
BC143918.1 - 1 246 9052436
BC143919.1 - 1 246

View Full Product Details