Gene Transcripts

Gene Symbol
TUBG2
Entrez Gene ID
27175
Gene Name
tubulin gamma 2
Gene Aliases
Chromosome Location
Chr. 17: 42659248 - 42667006 on Build GRCh38
UniGene ID
Hs.279669
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320509.1 NP_001307438.1 7 844
NM_016437.2 NP_057521.1 6 817
XM_011524614.2 XP_011522916.1 5 752
XM_017024453.1 XP_016879942.1 5 732
XM_017024454.1 XP_016879943.1 6 2003
XM_017024455.1 XP_016879944.1 6 831
XM_017024456.1 XP_016879945.1 6 2080
XM_017024457.1 XP_016879946.1 7 645
GenBank mRNA AF225971.1 - 6 810
AK022324.1 - 6 760
AK225134.1 - 6 776
BC009670.2 - 6 669 3891214
BC051890.1 - 6 760 5726689
BC108739.1 - 6 778 5242733

View Full Product Details