Gene Transcripts

Gene Symbol
AHDC1
Entrez Gene ID
27245
Gene Name
AT-hook DNA binding motif containing 1
Gene Aliases
MRD25
Chromosome Location
Chr. 1: 27534245 - 27604178 on Build GRCh38
UniGene ID
Hs.469280
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029882.3 NP_001025053.1 6 1532
XM_005245848.3 XP_005245905.1 7 1767
XM_005245849.3 XP_005245906.1 7 1872
XM_005245850.3 XP_005245907.1 8 1892
XM_005245851.3 XP_005245908.1 7 1764
XM_005245852.3 XP_005245909.1 6 1541
XM_011541255.1 XP_011539557.1 7 1446
XM_011541256.2 XP_011539558.1 7 1886
XM_011541257.2 XP_011539559.1 6 2641
GenBank mRNA BC064935.1 - 8 1467

View Full Product Details