Gene Transcripts
- Gene Symbol
- UBE2T
- Entrez Gene ID
- 29089
- Gene Name
- ubiquitin conjugating enzyme E2 T
- Gene Aliases
- FANCT, HSPC150, PIG50
- Chromosome Location
- Chr. 1: 202331657 - 202341966 on Build GRCh38
- UniGene ID
- Hs.5199
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001310326.1 |
NP_001297255.1
|
5, 6 |
528 |
|
NM_014176.3 |
NP_054895.1
|
5, 6 |
524 |
|
GenBank mRNA |
AB032931.1 |
- |
4, 5 |
430 |
|
AF160215.1 |
- |
5, 6 |
481 |
|
AF161499.1 |
- |
5, 6 |
514 |
|
AK000504.1 |
- |
5, 6 |
479 |
|
AY542309.1 |
- |
4, 5 |
389 |
|
BC004152.2 |
- |
5, 6 |
486 |
2821806 |
BC019284.1 |
- |
5, 6 |
486 |
2821806 |
BG776416.1 |
- |
5, 6 |
525 |
|
BM460257.1 |
- |
5, 6 |
501 |
|
BP244265.1 |
- |
5, 6 |
524 |
|
View Full Product Details