Gene Transcripts

Gene Symbol
UQCR10
Entrez Gene ID
29796
Gene Name
ubiquinol-cytochrome c reductase, complex III subunit X
Gene Aliases
HSPC051, HSPC119, HSPC151, QCR9, UCCR7.2, UCRC
Chromosome Location
Chr. 22: 29767369 - 29770413 on Build GRCh38
UniGene ID
Hs.284292
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003684.1 NP_001003684.1 2 419
NM_013387.3 NP_037519.2 2 360
GenBank mRNA AB028598.1 - 2 361
AF112217.1 - 2 349
AF161468.1 - 2 347
AF161500.1 - 2 417
AF161536.1 - 2 366
AK225973.1 - 2 345
BC005402.1 - 2 360 3961930
BC015971.1 - 2 354 4097106
BI754905.1 - 2 322
BU935406.1 - 419
CB999839.1 - 2 362
CD674029.1 - 2 227

View Full Product Details