Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001079514.2 | NP_001072982.1 | 4 | 1037 | |
NM_001288656.1 | NP_001275585.1 | 3 | 471 | ||
XM_005255277.3 | XP_005255334.1 | 4 | 1222 | ||
XM_005255278.3 | XP_005255335.1 | 4 | 2186 | ||
XM_005255279.3 | XP_005255336.1 | 4 | 1222 | ||
XM_011522465.1 | XP_011520767.1 | 4 | 1222 | ||
XM_011522467.1 | XP_011520769.1 | 4 | 1222 | ||
XM_017023189.1 | XP_016878678.1 | 4 | 1222 | ||
XM_017023190.1 | XP_016878679.1 | 4 | 1222 | ||
XM_017023192.1 | XP_016878681.1 | 4 | 1222 | ||
GenBank mRNA | AF108460.1 | - | 4 | 532 | |
AF108461.1 | - | 3 | 1119 | ||
AK056282.1 | - | 4 | 2186 | ||
AK300131.1 | - | 3 | 471 | ||
BC013750.1 | - | 4 | 757 |