Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013335.3 | NP_037467.2 | 9 | 897 | |
NM_205847.2 | NP_995319.1 | 9 | 1178 | ||
XM_005246483.3 | XP_005246540.1 | 10 | 912 | ||
XM_005246486.3 | XP_005246543.1 | 9 | 866 | ||
XM_011511032.2 | XP_011509334.1 | 10 | 881 | ||
XM_017003906.1 | XP_016859395.1 | 10 | 912 | ||
XM_017003907.1 | XP_016859396.1 | 10 | 881 | ||
XM_017003908.1 | XP_016859397.1 | 10 | 1187 | ||
XM_017003909.1 | XP_016859398.1 | 9 | 897 | ||
XM_017003910.1 | XP_016859399.1 | 9 | 866 | ||
XM_017003911.1 | XP_016859400.1 | 9 | 1172 | ||
XM_017003912.1 | XP_016859401.1 | 8 | 762 | ||
XM_017003913.1 | XP_016859402.1 | 8 | 731 | ||
XM_017003914.1 | XP_016859403.1 | 10 | 841 | ||
XM_017003915.1 | XP_016859404.1 | 10 | 1071 | ||
XM_017003916.1 | XP_016859405.1 | 9 | 826 | ||
XM_017003917.1 | XP_016859406.1 | 9 | 826 | ||
GenBank mRNA | AB209592.1 | - | 4 | 3640 | |
AF135422.2 | - | 8 | 876 | ||
AK000999.1 | - | 9 | 871 | ||
AK022578.1 | - | 9 | 1172 | ||
AK222951.1 | - | 9 | 861 | ||
AK290671.1 | - | 9 | 1174 | ||
BC007456.2 | - | 9 | 1166 | 2989461 |