Gene Transcripts

Gene Symbol
GMPPA
Entrez Gene ID
29926
Gene Name
GDP-mannose pyrophosphorylase A
Gene Aliases
AAMR
Chromosome Location
Chr. 2: 219498865 - 219506996 on Build GRCh38
UniGene ID
Hs.27059
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013335.3 NP_037467.2 6 563
NM_205847.2 NP_995319.1 6 844
XM_005246483.3 XP_005246540.1 6 563
XM_005246486.3 XP_005246543.1 6 532
XM_011511032.2 XP_011509334.1 6 532
XM_017003906.1 XP_016859395.1 6 563
XM_017003907.1 XP_016859396.1 6 532
XM_017003908.1 XP_016859397.1 6 838
XM_017003909.1 XP_016859398.1 6 563
XM_017003910.1 XP_016859399.1 6 532
XM_017003911.1 XP_016859400.1 6 838
XM_017003912.1 XP_016859401.1 6 563
XM_017003913.1 XP_016859402.1 6 532
XM_017003914.1 XP_016859403.1 6 492
XM_017003915.1 XP_016859404.1 6 722
XM_017003916.1 XP_016859405.1 6 492
XM_017003917.1 XP_016859406.1 6 492
GenBank mRNA AB209592.1 - 3 2197
AF135422.2 - 5 542
AK000999.1 - 6 537
AK022578.1 - 6 838
AK222951.1 - 6 527
AK290671.1 - 6 840
AK296193.1 - 5 790
BC007456.2 - 6 832 2989461
BP366441.1 - 6 502

View Full Product Details