Gene Transcripts

Gene Symbol
EHD2
Entrez Gene ID
30846
Gene Name
EH domain containing 2
Gene Aliases
PAST2
Chromosome Location
Chr. 19: 47713344 - 47743134 on Build GRCh38
UniGene ID
Hs.744963
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014601.3 NP_055416.2 5 1178
GenBank mRNA AF181263.1 - 4 966
AF454952.1 - 4 983
AK074701.1 - 5 1100
AK097126.1 - 5 505
AK097228.1 - 4 497
AK293429.1 - 1 27
AK296907.1 - 6 1070
AK298067.1 - 4 641
AK300512.1 - 2 129
AK315548.1 - 5 1102
AK316448.1 - 2 78
BC014445.1 - 5 1089 4908085

View Full Product Details