Gene Transcripts

Gene Symbol
CTAG2
Entrez Gene ID
30848
Gene Name
cancer/testis antigen 2
Gene Aliases
CAMEL, CT2, CT6.2, CT6.2a, CT6.2b, ESO2, LAGE-1, LAGE2B
Chromosome Location
Chr. X: 154651972 - 154653579 on Build GRCh38
UniGene ID
Hs.713312
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020994.4 NP_066274.2 2 947
NM_172377.4 NP_758965.2 3 718
GenBank mRNA AJ012833.1 - 3 623
AJ012834.1 - 3 707
AJ012835.1 - 2 938
AJ223040.1 - 2 947
AJ223041.1 - 3 706
AY679088.1 - 3 706
AY679089.1 - 3 706
BC002833.2 - 2 901 3638129

Gene Symbol
CTAG1A
Entrez Gene ID
246100
Gene Name
cancer/testis antigen 1A
Gene Aliases
CT6.1, ESO1, LAGE-2, LAGE2A, NY-ESO-1
Chromosome Location
Chr. X: 154585154 - 154586816 on Build GRCh38
UniGene ID
Hs.534310
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139250.1 NP_640343.1 3 707

Gene Symbol
CTAG1B
Entrez Gene ID
1485
Gene Name
cancer/testis antigen 1B
Gene Aliases
CT6.1, CTAG, CTAG1, ESO1, LAGE-2, LAGE2B, NY-ESO-1
Chromosome Location
Chr. X: 154617609 - 154619271 on Build GRCh38
UniGene ID
Hs.534310
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001327.2 NP_001318.1 3 707
GenBank mRNA AF038567.1 - 3 742
AJ003149.1 - 3 704
AJ275978.1 - 2 815
U87459.1 - 3 707

View Full Product Details