Gene Transcripts

Gene Symbol
SMARCAL1
Entrez Gene ID
50485
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a like 1
Gene Aliases
HARP, HHARP
Chromosome Location
Chr. 2: 216412414 - 216483053 on Build GRCh38
UniGene ID
Hs.516674
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127207.1 NP_001120679.1 9 1735
NM_014140.3 NP_054859.2 9 1921
XM_005246631.2 XP_005246688.1 9 1859
XM_005246632.1 XP_005246689.1 9 1911
XM_017004228.1 XP_016859717.1 9 1724
GenBank mRNA AF082179.1 - 8 1691
AF432223.1 - 9 1731
AK000117.1 - 5 591
AK023780.1 - 9 1862
AL122076.1 - 7 1155
BC016482.1 - 8 1682 4341673
BC029994.1 - 7 1183
BC043341.1 - 9 1708 6058575

View Full Product Details