Gene Transcripts

Gene Symbol
SYCP3
Entrez Gene ID
50511
Gene Name
synaptonemal complex protein 3
Gene Aliases
COR1, RPRGL4, SCP3, SPGF4
Chromosome Location
Chr. 12: 101728648 - 101739722 on Build GRCh38
UniGene ID
Hs.506504
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177948.1 NP_001171419.1 8 705
NM_001177949.1 NP_001171420.1 8 720
NM_153694.4 NP_710161.1 8 716
XM_005268922.4 XP_005268979.3 8 966
XM_005268924.1 XP_005268981.1 8 704
XM_005268925.1 XP_005268982.1 8 709
XM_005268926.3 XP_005268983.1 8 688
XM_005268927.2 XP_005268984.1 8 677
XM_011538421.2 XP_011536723.2 8 888
XM_017019368.1 XP_016874857.1 8 963
GenBank mRNA AF492003.1 - 8 661
AF517774.1 - 8 659
AK125930.1 - 5 3278
BC062662.1 - 8 711 6616222
BG201028.1 - 2 131
BG202454.1 - 2 135

View Full Product Details