Gene Transcripts
- Gene Symbol
- SYCP3
- Entrez Gene ID
- 50511
- Gene Name
- synaptonemal complex protein 3
- Gene Aliases
- COR1, RPRGL4, SCP3, SPGF4
- Chromosome Location
- Chr. 12: 101728648 - 101739722 on Build GRCh38
- UniGene ID
- Hs.506504
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001177948.1 |
NP_001171419.1
|
2, 3 |
266 |
|
NM_001177949.1 |
NP_001171420.1
|
2, 3 |
281 |
|
NM_153694.4 |
NP_710161.1
|
2, 3 |
277 |
|
XM_005268922.4 |
XP_005268979.3
|
2, 3 |
527 |
|
XM_005268924.1 |
XP_005268981.1
|
2, 3 |
265 |
|
XM_005268925.1 |
XP_005268982.1
|
2, 3 |
270 |
|
XM_005268926.3 |
XP_005268983.1
|
2, 3 |
249 |
|
XM_005268927.2 |
XP_005268984.1
|
2, 3 |
238 |
|
XM_011538421.2 |
XP_011536723.2
|
2, 3 |
449 |
|
XM_017019368.1 |
XP_016874857.1
|
2, 3 |
524 |
|
GenBank mRNA |
AF492003.1 |
- |
2, 3 |
222 |
|
AF517774.1 |
- |
2, 3 |
220 |
|
AK125930.1 |
- |
2 |
772 |
|
BC062662.1 |
- |
2, 3 |
272 |
6616222 |
BQ428625.1 |
- |
2, 3 |
282 |
|
View Full Product Details