Gene Transcripts
- Gene Symbol
- TAS2R3
- Entrez Gene ID
- 50831
- Gene Name
- taste 2 receptor member 3
- Gene Aliases
- T2R3
- Chromosome Location
- Chr. 7: 141764097 - 141765197 on Build GRCh38
- UniGene ID
- Hs.676011
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016943.2 |
NP_058639.1
|
1 |
876 |
|
View Full Product Details