Gene Transcripts

Gene Symbol
TAS2R10
Entrez Gene ID
50839
Gene Name
taste 2 receptor member 10
Gene Aliases
T2R10, TRB2
Chromosome Location
Chr. 12: 10825346 - 10826269 on Build GRCh38
UniGene ID
Hs.533756
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023921.1 NP_076410.1 1 843
GenBank mRNA BC069089.1 - 1 843
BC101762.1 - 1 932 8069268
BC101764.1 - 1 932 8069270

View Full Product Details