Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001197026.1 | NP_001183955.1 | 14 | 1814 | |
XM_011515591.2 | XP_011513893.1 | 15 | 1879 | ||
XM_011515596.2 | XP_011513898.1 | 13 | 1330 | ||
XM_017012746.1 | XP_016868235.1 | 14 | 1821 | ||
XM_017012747.1 | XP_016868236.1 | 14 | 1854 | ||
GenBank mRNA | AF308300.1 | - | 14 | 1491 | |
AK294857.1 | - | 14 | 1763 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_037144.1 | 3 | 1601 | ||
GenBank mRNA | AF103731.1 | - | 3 | 1601 | |
AF380162.1 | - | 1 | 1430 | ||
AK024218.1 | - | 1 | 576 | ||
BC028914.1 | - | 1 | 1442 | ||
BC063575.1 | - | 3 | 1482 | 3881751 |