Gene Transcripts

Gene Symbol
SEPT7
Entrez Gene ID
989
Gene Name
septin 7
Gene Aliases
CDC10, CDC3, NBLA02942, SEPT7A
Chromosome Location
Chr. 7: 35800986 - 35915763 on Build GRCh38
UniGene ID
Hs.191346
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011553.3 NP_001011553.2 5 613
NM_001242956.1 NP_001229885.1 4 508
NM_001788.5 NP_001779.3 6 616
XM_006715806.3 XP_006715869.1 6 564
XM_011515656.2 XP_011513958.1 6 564
XM_011515661.2 XP_011513963.1 6 564
XM_017012855.1 XP_016868344.1 6 548
XM_017012856.1 XP_016868345.1 6 556
XM_017012857.1 XP_016868346.1 6 553
XM_017012858.1 XP_016868347.1 7 478
XM_017012859.1 XP_016868348.1 6 475
XM_017012860.1 XP_016868349.1 7 472
XM_017012861.1 XP_016868350.1 7 469
XM_017012862.1 XP_016868351.1 5 379
XM_017012863.1 XP_016868352.1 6 922
GenBank mRNA AB073389.1 - 6 383
AB209677.1 - 6 351
AK223225.1 - 6 383
AK223241.1 - 6 383
AK290545.1 - 5 454
AK297883.1 - 4 275
AK298453.1 - 6 450
AK300601.1 - 7 329
BC025987.2 - 5 380 4823810
BC067264.1 - 5 359 6503495
BC093640.1 - 6 382 7939485
BC093642.1 - 6 382 7939487
BC107718.1 - 5 561
BI669760.1 - 6 594
BQ229116.1 - 5 544
BQ232357.1 - 6 358
BX571755.1 - 5 381
DQ232879.1 - 5 364
KF305319.1 - 7 448
S72008.1 - 6 383

View Full Product Details