Gene Transcripts

Gene Symbol
A4GNT
Entrez Gene ID
51146
Gene Name
alpha-1,4-N-acetylglucosaminyltransferase
Gene Aliases
alpha4GnT
Chromosome Location
Chr. 3: 138123718 - 138133310 on Build GRCh38
UniGene ID
Hs.278960
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016161.2 NP_057245.1 3 825
XM_017006543.1 XP_016862032.1 3 1376
XM_017006544.1 XP_016862033.1 3 946
GenBank mRNA AF141315.1 - 3 803
AK225668.1 - 3 825
BC119639.1 - 3 720 40115688
BC119640.2 - 3 720 40115689

View Full Product Details