Gene Transcripts

Gene Symbol
CDC25B
Entrez Gene ID
994
Gene Name
cell division cycle 25B
Gene Aliases
Chromosome Location
Chr. 20: 3786772 - 3806121 on Build GRCh38
UniGene ID
Hs.153752
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287516.1 NP_001274445.1 13 1462
NM_001287517.1 NP_001274446.1 13 1420
NM_001287518.1 NP_001274447.1 12 1381
NM_001287519.1 NP_001274448.1 13 1193
NM_001287520.1 NP_001274449.1 13 1235
NM_001287522.1 NP_001274451.1 12 1112
NM_001287524.1 NP_001274453.1 9 1017
NM_004358.4 NP_004349.1 13 2045
NM_021872.3 NP_068658.1 12 1964
NM_021873.3 NP_068659.1 13 2087
NR_136335.1 14 1314
NR_136336.1 14 1541
GenBank mRNA AK092713.1 - 13 1235
AK295573.1 - 13 1304
AK299028.1 - 12 1188
AK299192.1 - 13 1238
BC006395.2 - 13 1368 4135465
BC009953.2 - 13 1448 4130804
BC051711.1 - 13 1992 5301205
BX640836.1 - 9 1009
BX647988.1 - 13 1150
FR695900.1 - 13 1235
FR695901.1 - 12 1112
M81934.1 - 13 1324
S78187.1 - 13 1492
Z68092.1 - 12 1272

View Full Product Details