Gene Transcripts
- Gene Symbol
- CERCAM
- Entrez Gene ID
- 51148
- Gene Name
- cerebral endothelial cell adhesion molecule
- Gene Aliases
- CEECAM1, GLT25D3
- Chromosome Location
- Chr. 9: 128405993 - 128437351 on Build GRCh38
- UniGene ID
- Hs.495230
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286760.1 |
NP_001273689.1
|
3, 4 |
346 |
|
NM_016174.4 |
NP_057258.3
|
3, 4 |
822 |
|
XM_005252035.1 |
XP_005252092.1
|
5, 6 |
1732 |
|
XM_011518762.1 |
XP_011517064.1
|
6, 7 |
1889 |
|
XM_011518763.2 |
XP_011517065.1
|
3, 4 |
1440 |
|
XM_017014794.1 |
XP_016870283.1
|
7, 8 |
1631 |
|
GenBank mRNA |
AB040935.1 |
- |
4, 5 |
3335 |
|
AF177203.1 |
- |
2, 3 |
190 |
|
AK074519.1 |
- |
2, 3 |
258 |
|
AK074523.1 |
- |
2, 3 |
232 |
|
AK297929.1 |
- |
2, 3 |
318 |
|
AK314294.1 |
- |
3, 4 |
393 |
|
BC098432.1 |
- |
3, 4 |
420 |
6672104 |
BC108698.1 |
- |
3, 4 |
433 |
|
BC119698.1 |
- |
3, 4 |
295 |
40116832 |
BC119699.1 |
- |
3, 4 |
295 |
40116834 |
BX358794.2 |
- |
3, 4 |
471 |
|
View Full Product Details