Gene Transcripts

Gene Symbol
AADAT
Entrez Gene ID
51166
Gene Name
aminoadipate aminotransferase
Gene Aliases
KAT2, KATII, KYAT2
Chromosome Location
Chr. 4: 170060222 - 170091699 on Build GRCh38
UniGene ID
Hs.529735
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286682.1 NP_001273611.1 9 979
NM_001286683.1 NP_001273612.1 9 1052
NM_016228.3 NP_057312.1 8 1185
NM_182662.1 NP_872603.1 9 967
XM_006714231.2 XP_006714294.1 10 1109
XM_011532020.2 XP_011530322.1 6 742
GenBank mRNA AF481738.1 - 8 1186
AK055952.1 - 9 967
AK314729.1 - 9 1054
BC031068.1 - 9 972 5286180

View Full Product Details