Gene Transcripts

Gene Symbol
HSPA14
Entrez Gene ID
51182
Gene Name
heat shock protein family A (Hsp70) member 14
Gene Aliases
HSP70-4, HSP70L1
Chromosome Location
Chr. 10: 14838160 - 14871741 on Build GRCh38
UniGene ID
Hs.534169
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016299.3 NP_057383.2 11 1352
GenBank mRNA AF112210.1 - 11 1171
AF143723.1 - 11 1161
AK225265.1 - 11 1196
AK225651.1 - 11 1196
AK226065.1 - 4 4025
AK292323.1 - 11 1160
BC119690.1 - 11 1171 40116697

View Full Product Details