Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318221.1 | NP_001305150.1 | 3 | 262 | |
NM_001318222.1 | NP_001305151.1 | 3 | 193 | ||
NM_016633.3 | NP_057717.1 | 3 | 266 | ||
GenBank mRNA | AF112995.1 | - | 3 | 297 | |
AF147435.1 | - | 3 | 225 | ||
AF208865.1 | - | 3 | 222 | ||
AF364517.1 | - | 3 | 222 | ||
BC035842.1 | - | 3 | 207 | 5748040 | |
BG940214.1 | - | 3 | 221 | ||
BG943964.1 | - | 3 | 215 | ||
H94920.1 | - | 3 | 255 |