Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001278625.1 | NP_001265554.1 | 9 | 1504 | |
NM_001278626.1 | NP_001265555.1 | 9 | 1317 | ||
NM_001278627.1 | NP_001265556.1 | 9 | 1227 | ||
NM_001278628.1 | NP_001265557.1 | 8 | 1175 | ||
NM_016652.5 | NP_057736.4 | 9 | 1540 | ||
GenBank mRNA | AB016840.1 | - | 8 | 1137 | |
AF111802.1 | - | 8 | 1131 | ||
AF255443.2 | - | 9 | 1504 | ||
AF318302.1 | - | 8 | 1122 | ||
AF318303.1 | - | 9 | 1540 | ||
AF318304.1 | - | 9 | 1317 | ||
AF318305.1 | - | 9 | 1227 | ||
AK022908.1 | - | 7 | 852 | ||
AK023246.1 | - | 5 | 687 | ||
AK023728.1 | - | 4 | 501 | ||
AK292799.1 | - | 8 | 1045 | ||
AL832435.1 | - | 8 | 1149 |