Gene Transcripts

Gene Symbol
POMP
Entrez Gene ID
51371
Gene Name
proteasome maturation protein
Gene Aliases
C13orf12, HSPC014, PNAS-110, UMP1
Chromosome Location
Chr. 13: 28659004 - 28678957 on Build GRCh38
UniGene ID
Hs.268742
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015932.5 NP_057016.1 5 479
GenBank mRNA AF077200.1 - 5 380
AF125097.1 - 5 379
AF262975.1 - 5 323
AF275807.1 - 5 328
AK312118.1 - 5 350
BC003390.1 - 5 342 3446986
BC014334.1 - 5 373 4045091
EF535527.1 - 5 311
GU045555.1 - 5 479
GU045556.1 - 5 478

View Full Product Details