Gene Transcripts

Gene Symbol
TMA7
Entrez Gene ID
51372
Gene Name
translation machinery associated 7 homolog
Gene Aliases
CCDC72, HSPC016
Chromosome Location
Chr. 3: 48440276 - 48444129 on Build GRCh38
UniGene ID
Hs.356440
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF161448.1 - 3 400
AK126594.1 - 4 2311
AL712164.1 - 4 428
AY794023.1 - 4 435

View Full Product Details