Gene Transcripts
- Gene Symbol
- ANAPC5
- Entrez Gene ID
- 51433
- Gene Name
- anaphase promoting complex subunit 5
- Gene Aliases
- APC5
- Chromosome Location
- Chr. 12: 121308245 - 121354209 on Build GRCh38
- UniGene ID
- Hs.7101
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001137559.1 |
NP_001131031.1
|
13, 14 |
1506 |
|
NM_016237.4 |
NP_057321.2
|
13, 14 |
1767 |
|
XM_005253900.3 |
XP_005253957.1
|
13, 14 |
1730 |
|
XM_017019423.1 |
XP_016874912.1
|
12, 13 |
1403 |
|
XM_017019424.1 |
XP_016874913.1
|
12, 13 |
1364 |
|
GenBank mRNA |
AF191339.1 |
- |
13, 14 |
1645 |
|
AK025614.1 |
- |
13, 14 |
1506 |
|
AK054779.1 |
- |
3, 4 |
1343 |
|
AK294139.1 |
- |
6, 7 |
763 |
|
AK301197.1 |
- |
5, 6 |
997 |
|
AK315451.1 |
- |
13, 14 |
1712 |
|
BC001081.2 |
- |
13, 14 |
1700 |
2823401 |
BC001950.1 |
- |
13, 14 |
1700 |
2823401 |
BC003095.1 |
- |
8, 9 |
989 |
|
BC006301.2 |
- |
13, 14 |
1704 |
4100632 |
BC034243.1 |
- |
12, 13 |
1407 |
|
BC045686.1 |
- |
6, 7 |
1855 |
|
BX537687.1 |
- |
8, 9 |
1276 |
|
View Full Product Details