Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014306.4 | NP_055121.1 | 9 | 1232 | |
GenBank mRNA | AF155658.1 | - | 9 | 1180 | |
AF161466.1 | - | 9 | 1191 | ||
AK297826.1 | - | 9 | 1109 | ||
AK312503.1 | - | 9 | 1192 | ||
AL050255.1 | - | 9 | 1176 | ||
AL137272.1 | - | 2 | 178 | ||
BC000151.1 | - | 9 | 1170 | 2984824 | |
BC002970.1 | - | 9 | 1170 | 3533883 | |
BC010308.1 | - | 9 | 1182 | 3451576 | |
BC016707.1 | - | 9 | 1197 | 4102148 | |
CR456450.1 | - | 9 | 1147 | ||
CR457175.1 | - | 9 | 1101 |