Gene Transcripts

Gene Symbol
UFC1
Entrez Gene ID
51506
Gene Name
ubiquitin-fold modifier conjugating enzyme 1
Gene Aliases
HSPC155
Chromosome Location
Chr. 1: 161132754 - 161158856 on Build GRCh38
UniGene ID
Hs.301412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016406.3 NP_057490.2 2, 3 442
XM_005245254.1 XP_005245311.1 2, 3 394
XM_017001450.1 XP_016856939.1 3, 4 287
XM_017001451.1 XP_016856940.1 3, 4 551
GenBank mRNA AB154405.1 - 2, 3 188
AF151884.1 - 2, 3 409
AF161504.1 - 2, 3 428
AK292776.1 - 2, 3 394
BC005187.1 - 2, 3 434 3961569
CF128121.1 - 2, 3 441

View Full Product Details