Gene Transcripts
- Gene Symbol
- CDK8
- Entrez Gene ID
- 1024
- Gene Name
- cyclin dependent kinase 8
- Gene Aliases
- K35
- Chromosome Location
- Chr. 13: 26254104 - 26405238 on Build GRCh38
- UniGene ID
- Hs.382306
- Species
- Homo sapiens
- Gene Symbol
- ADRA1B
- Entrez Gene ID
- 147
- Gene Name
- adrenoceptor alpha 1B
- Gene Aliases
- ADRA1, ALPHA1BAR
- Chromosome Location
- Chr. 5: 159916733 - 159989205 on Build GRCh38
- UniGene ID
- Hs.368632
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK309989.1 |
- |
|
1320 |
|
View Full Product Details