Gene Transcripts

Gene Symbol
LIPT1
Entrez Gene ID
51601
Gene Name
lipoyltransferase 1
Gene Aliases
LIPT1D
Chromosome Location
Chr. 2: 99154955 - 99163157 on Build GRCh38
UniGene ID
Hs.719172
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204830.1 NP_001191759.1 3 583
NM_015929.3 NP_057013.1 3 385
NM_145197.2 NP_660198.1 4 513
NM_145198.2 NP_660199.1 4 501
NM_145199.2 NP_660200.1 2 259
NR_037935.1 4 646
NR_037936.1 3 322
GenBank mRNA AB017566.1 - 3 385
BC007001.1 - 3 305 3961227
BC009772.1 - 3 326 4091817
BC070145.1 - 3 363 4671058
BG717409.1 - 3 561
BQ422048.1 - 3 294
BQ432795.1 - 4 600

View Full Product Details