Gene Transcripts
- Gene Symbol
- STYXL1
- Entrez Gene ID
- 51657
- Gene Name
- serine/threonine/tyrosine interacting-like 1
- Gene Aliases
- DUSP24, MK-STYX, MKSTYX
- Chromosome Location
- Chr. 7: 75995820 - 76048709 on Build GRCh38
- UniGene ID
- Hs.11615
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001317785.1 |
NP_001304714.1
|
7, 8 |
1040 |
|
NM_001317786.1 |
NP_001304715.1
|
7, 8 |
890 |
|
NM_001317787.1 |
NP_001304716.1
|
5, 6 |
599 |
|
NM_001317789.1 |
NP_001304718.1
|
5, 6 |
549 |
|
NM_016086.2 |
NP_057170.1
|
7, 8 |
1037 |
|
XM_011516293.2 |
XP_011514595.1
|
7, 8 |
883 |
|
XM_011516300.1 |
XP_011514602.1
|
6, 7 |
693 |
|
XM_017012296.1 |
XP_016867785.1
|
8, 9 |
997 |
|
XM_017012300.1 |
XP_016867789.1
|
8, 9 |
917 |
|
XM_017012301.1 |
XP_016867790.1
|
6, 7 |
737 |
|
XM_017012302.1 |
XP_016867791.1
|
6, 7 |
740 |
|
GenBank mRNA |
AF069762.1 |
- |
7, 8 |
1033 |
|
AF188202.1 |
- |
3, 4 |
271 |
|
AF188204.1 |
- |
4, 5 |
414 |
|
AK308912.1 |
- |
7, 8 |
864 |
|
AL533027.3 |
- |
7, 8 |
838 |
|
BC024035.2 |
- |
7, 8 |
1000 |
2988900 |
BX537896.1 |
- |
5, 6 |
579 |
|
View Full Product Details