Gene Transcripts

Gene Symbol
ZFR
Entrez Gene ID
51663
Gene Name
zinc finger RNA binding protein
Gene Aliases
SPG71, ZFR1
Chromosome Location
Chr. 5: 32354350 - 32444738 on Build GRCh38
UniGene ID
Hs.435231
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016107.3 NP_057191.2 10 1835
GenBank mRNA AJ314790.1 - 10 1788
AK292214.1 - 10 1835
BC030540.1 - 10 1807
BC051893.1 - 3 426
BC062986.1 - 3 351
BC137084.1 - 10 1799 9021081
BC137085.1 - 10 1799 9021082

View Full Product Details