Gene Transcripts

Gene Symbol
LSM7
Entrez Gene ID
51690
Gene Name
LSM7 homolog, U6 small nuclear RNA and mRNA degradation associated
Gene Aliases
YNL147W
Chromosome Location
Chr. 19: 2321521 - 2328615 on Build GRCh38
UniGene ID
Hs.512610
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016199.2 NP_057283.1 3, 4 215
XM_017026869.1 XP_016882358.1 4, 5 358
GenBank mRNA AF182293.1 - 2, 3 175
BC018621.1 - 2, 3 168 4855162
BI553911.1 - 3, 4 219

View Full Product Details