Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164831.2 | NP_001158303.1 | 4 | 355 | |
NM_001164832.2 | NP_001158304.1 | 5 | 704 | ||
NM_001164836.2 | NP_001158308.1 | 6 | 780 | ||
NM_001164837.2 | NP_001158309.1 | 5 | 683 | ||
NM_022003.3 | NP_071286.1 | 4 | 607 | ||
GenBank mRNA | AK289629.1 | - | 4 | 547 | |
AK290757.1 | - | 6 | 482 | ||
AK311736.1 | - | 4 | 219 | ||
AL035858.1 | - | 4 | 231 | ||
AL136699.1 | - | 4 | 193 | ||
AL832811.1 | - | 5 | 300 | ||
AY358976.1 | - | 4 | 217 | ||
BC018652.2 | - | 6 | 390 | 2822864 | |
BC093040.1 | - | 4 | 217 | 6526843 | |
BT006717.1 | - | 3 | 162 | ||
BX460202.2 | - | 5 | 352 | ||
CR533457.1 | - | 3 | 162 | ||
DA001721.1 | - | 4 | 195 | ||
DA056858.1 | - | 4 | 353 | ||
DA086584.1 | - | 4 | 299 | ||
DA131017.1 | - | 4 | 311 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001204268.1 | NP_001191197.1 | 4 | 607 | |
NM_001243598.2 | NP_001230527.1 | 4 | 607 | ||
GenBank mRNA | CN310658.1 | - | 4 | 242 | |
FY211982.1 | - | 4 | 204 | ||
FY214031.1 | - | 4 | 204 |