Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001006114.2 | NP_001006114.1 | 2 | 285 | |
NM_001314022.1 | NP_001300951.1 | 2 | 607 | ||
NM_001314025.1 | NP_001300954.1 | 2 | 285 | ||
NM_001314026.1 | NP_001300955.1 | 2 | 607 | ||
NM_058181.2 | NP_478061.1 | 2 | 607 | ||
XM_005261156.3 | XP_005261213.2 | 2 | 559 | ||
XM_011529629.2 | XP_011527931.1 | 2 | 255 | ||
XM_011529630.2 | XP_011527932.1 | 2 | 286 | ||
XM_011529631.2 | XP_011527933.1 | 2 | 255 | ||
XM_011529633.2 | XP_011527935.1 | 2 | 255 | ||
XM_011529634.2 | XP_011527936.2 | 2 | 559 | ||
XM_011529635.2 | XP_011527937.1 | 2 | 286 | ||
XM_017028394.1 | XP_016883883.1 | 2 | 559 | ||
XM_017028396.1 | XP_016883885.1 | 302 | |||
GenBank mRNA | AK128875.1 | - | 2 | 219 | |
AK294975.1 | - | 2 | 266 | ||
AY040873.1 | - | 2 | 553 | ||
AY040874.1 | - | 2 | 553 | ||
BC022828.1 | - | 2 | 226 | 3928883 | |
BC119770.1 | - | 2 | 231 | 40108760 | |
BC122865.1 | - | 2 | 231 | 40108759 | |
BC131553.1 | - | 2 | 231 | 40089560 | |
BG473705.1 | - | 1 | 191 | ||
BM796288.1 | - | 2 | 278 |