Gene Transcripts

Gene Symbol
ETAA1
Entrez Gene ID
54465
Gene Name
Ewing tumor associated antigen 1
Gene Aliases
ETAA16
Chromosome Location
Chr. 2: 67397310 - 67410411 on Build GRCh38
UniGene ID
Hs.353022
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019002.3 NP_061875.2 5 1848
XM_017004376.1 XP_016859865.1 5 1841
XM_017004377.1 XP_016859866.1 5 1841
GenBank mRNA AJ242682.2 - 5 1853
AK025155.1 - 4 1381
AK026300.1 - 1 294
BC033075.1 - 5 1820
BC040001.1 - 5 1780 5556133

View Full Product Details