Gene Transcripts

Gene Symbol
TOLLIP
Entrez Gene ID
54472
Gene Name
toll interacting protein
Gene Aliases
IL-1RAcPIP
Chromosome Location
Chr. 11: 1274368 - 1309666 on Build GRCh38
UniGene ID
Hs.368527
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318512.1 NP_001305441.1 5 1240
NM_001318514.1 NP_001305443.1 6 1400
NM_001318515.1 NP_001305444.1 3 963
NM_001318516.1 NP_001305445.1 5 1207
NM_019009.3 NP_061882.2 6 1390
XM_017017931.1 XP_016873420.1 4 1297
GenBank mRNA AB209541.1 - 7 1095
AK022871.1 - 6 1297
AK090850.1 - 5 1201
AK096693.1 - 5 1164
AK127123.1 - 6 1353
AL136835.1 - 6 1337
BC004420.1 - 6 1350 3356339
BC012057.1 - 6 1315 4647828
BC018272.1 - 6 1307 3452585
BX648136.1 - 1 1667

View Full Product Details