Gene Transcripts

Gene Symbol
MARCH5
Entrez Gene ID
54708
Gene Name
membrane associated ring-CH-type finger 5
Gene Aliases
MARCH-V, MITOL, RNF153
Chromosome Location
Chr. 10: 92291163 - 92353964 on Build GRCh38
UniGene ID
Hs.573490
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017824.4 NP_060294.1 3 606
XM_011539886.2 XP_011538188.1 4 754
GenBank mRNA AB191202.1 - 3 362
AK000452.1 - 3 608
AK294799.1 - 3 680
BC015480.1 - 3 566 3905766

View Full Product Details