Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017515.4 | NP_059985.2 | 3 | 790 | |
GenBank mRNA | AK023080.1 | - | 3 | 387 | |
AK128062.1 | - | 6 | 959 | ||
AK300582.1 | - | 2 | 225 | ||
AL833969.1 | - | 3 | 393 | ||
BC039195.2 | - | 3 | 530 | 3847013 | |
BC048302.1 | - | 3 | 396 | 5499821 | |
X99961.1 | - | 3 | 333 |