Gene Transcripts
- Gene Symbol
- HYDIN
- Entrez Gene ID
- 54768
- Gene Name
- HYDIN, axonemal central pair apparatus protein
- Gene Aliases
- CILD5, HYDIN1, HYDIN2, PPP1R31
- Chromosome Location
- Chr. 16: 70802084 - 71230722 on Build GRCh38
- UniGene ID
- Hs.351914
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001198542.1 |
NP_001185471.1
|
11, 12 |
1570 |
|
NM_001198543.1 |
NP_001185472.1
|
11, 12 |
1540 |
|
NM_001270974.2 |
NP_001257903.1
|
11, 12 |
1638 |
|
NM_017558.4 |
NP_060028.2
|
11, 12 |
1638 |
|
XM_006721206.2 |
XP_006721269.1
|
11, 12 |
1532 |
|
XM_011523146.2 |
XP_011521448.1
|
11, 12 |
1565 |
|
XM_011523147.1 |
XP_011521449.1
|
11, 12 |
1532 |
|
XM_011523148.1 |
XP_011521450.1
|
11, 12 |
1605 |
|
XM_011523151.1 |
XP_011521453.1
|
11, 12 |
1564 |
|
XM_017023346.1 |
XP_016878835.1
|
11, 12 |
1810 |
|
GenBank mRNA |
AK022933.1 |
- |
11, 12 |
1572 |
|
AK299016.1 |
- |
11, 12 |
1527 |
|
AK299348.1 |
- |
11, 12 |
1570 |
|
AK308296.1 |
- |
9, 10 |
1157 |
|
BC028351.1 |
- |
11, 12 |
1519 |
|
- Gene Symbol
- HYDIN2
- Entrez Gene ID
- 100288805
- Gene Name
- HYDIN2, axonemal central pair apparatus protein (pseudogene)
- Gene Aliases
- HYDINP1
- Chromosome Location
- Chr. 1: 146547489 - 146822034 on Build GRCh38
- UniGene ID
- Hs.648712
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_103556.1 |
|
6, 7 |
818 |
|
GenBank mRNA |
AL833826.1 |
- |
6, 7 |
818 |
|
AL834340.1 |
- |
3, 4 |
308 |
|
View Full Product Details