Gene Transcripts

Gene Symbol
GID8
Entrez Gene ID
54994
Gene Name
GID complex subunit 8 homolog
Gene Aliases
C20orf11, TWA1
Chromosome Location
Chr. 20: 62938089 - 62948475 on Build GRCh38
UniGene ID
Hs.353013
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017896.2 NP_060366.1 5 720
GenBank mRNA AK000609.1 - 5 656
BC032120.2 - 5 690 4137113

View Full Product Details