Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001317338.1 | NP_001304267.1 | 5, 6 | 1106 | |
NM_017898.4 | NP_060368.2 | 5, 6 | 1106 | ||
XM_006711407.2 | XP_006711470.1 | 4, 5 | 1662 | ||
XM_011509684.1 | XP_011507986.1 | 5, 6 | 630 | ||
GenBank mRNA | AK000612.1 | - | 3, 4 | 245 | |
BC010366.1 | - | 4, 5 | 440 | ||
BC011973.1 | - | 5, 6 | 1018 | 3458649 | |
BC015829.2 | - | 3, 4 | 376 | ||
BC016859.1 | - | 5, 6 | 1008 | 3849257 |